Human recombinant IFN 1b, INF beta 1b (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized Interferon-beta 1b although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution IFNB 1b should be stored at 4 °C between 2-7 days and for future use below −18 °C. Please prevent freeze/thaw cycles.
- Biological activity:The specific activity as determined in viral resistance assay (human "Wish" cell line and VSV virus or the monkey VERO cell line with EMCV virus) was found to be 32,000,000 IU/mg.
- Protein synonyms:Leukocyte interferon
- Protein/peptide name:IFN 1b, INF beta 1b
- Purity:Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized Interferon beta-1b in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be Ser-Tyr-Asn-Leu-Leu.
- Formulation:Lyophilized from 1mg/ml solution containing 50mg Human Albumin 50mg dextrose.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together