Order Entry
Belgium
ContactUsLinkComponent
Treponema p15
Treponema p15
Supplier:  Biorbyt
Certificates
Treponema p15
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    Treponema p15
  • Purity:
    Treponema Pallidum protein is >95% pure as determined by 10% PAGE (coomassie staining). Treponema Pallidum protein was purified by proprietary chromatographic technique.
  • Formulation:
    70 mM Tris-HCl pH-8, 84mM NaCl, 14mM Glutathione, 30% Glycerol 0.2% Sarcosil.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together