Order Entry
Belgium
ContactUsLinkComponent
Toxoplasma ROP4 RH2
Toxoplasma ROP4 RH2
Supplier:  Biorbyt
Certificates
Toxoplasma ROP4 RH2
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    Toxoplasma ROP4 RH2
  • Purity:
    Protein is >95% pure as determined by 10% PAGE (coomassie staining). Toxoplasma protein was purified by proprietary chromatographic technique.
  • Formulation:
    25mM Tris-HCl pH 8.0, 5mM Glutathione, 1.5M urea and 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together