Order Entry
Belgium
ContactUsLinkComponent
Encephalitis NS3
Encephalitis NS3
Supplier:  Biorbyt
Certificates
Encephalitis NS3
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    Encephalitis NS3
  • Purity:
    Encephalitis protein is >95% pure as determined by 10% PAGE (coomassie staining). Encephalitis protein was purified by proprietary chromatographic technique.
  • Formulation:
    20 mM MES pH 6.5, 8M urea, 200 mM NaCl and 0.05% Tween-20.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together