Order Entry
Belgium
ContactUsLinkComponent
SARS nucleocaspid
SARS nucleocaspid
Supplier:  Biorbyt
Certificates
SARS nucleocaspid
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month at 4 °C.
  • Protein/peptide name:
    SARS Nucleocaspid
  • Purity:
    SARS Core protein is >95% pure as determined by 10% PAGE (coomassie staining). The SARS Core protein was purified by proprietary chromatographic technique.
  • Formulation:
    50mM Tris-HCl, 60mM NaCl and 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together