Order Entry
Belgium
ContactUsLinkComponent
M. pneumoniae (from E. coli)
M. pneumoniae (from E. coli)
Supplier:  Biorbyt
Certificates
M. pneumoniae (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Storage conditions:
    Two years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    M. pneumoniae
  • Purity:
    Greater than 95% as determined by 10% PAGE (Coomassie staining). The recombinant fusion protein was purified by GSH affinity chromatography technique.
  • Formulation:
    Mycoplasma Pneumoniae P1-C is formulated in 1x PBS and 0.5mM EDTA.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together