rHSV-2 gG
: Biorbyt
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:rHSV-2 gG
- Purity:HSV-2 gG protein is >95% pure as determined by 10% PAGE (coomassie staining). HSV-2 gG protein was purified by proprietary chromatographic technique.
- Formulation:0.1% SDS, 100mM NaCl, 50% glycerol.
- Tested applications:SDS-PAGE
Frequently Bought Together