Order Entry
Belgium
ContactUsLinkComponent
HIV-1 gp41 Long (from E. coli)
HIV-1 gp41 Long (from E. coli)
Supplier:  Biorbyt
Certificates
HIV-1 gp41 Long (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Storage conditions:
    HIV-1 gp41 Long is shipped at ambient temperature. Upon arrival, Store at −20 °C. Stable for Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    HIV-1 gp41 Long
  • Purity:
    Greater than 95.0% as determined by HPLC analysis SDS-PAGE.
  • Formulation:
    10mM Na2CO3, 10mM Na3EDTA, 10mM Mbeta mercaptoethanol, 0.05% tween-20.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together