Order Entry
Belgium
ContactUsLinkComponent
HCV genotype-2a
HCV genotype-2a
Supplier:  Biorbyt
Certificates
HCV genotype-2a
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    HCV Genotype-2a
  • Purity:
    HCV Core Genotype-2a protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-2a protein was purified by proprietary chromatographic technique.
  • Formulation:
    25mM Tris-HCl, pH 8.0, 1.5M Urea, 0.2% Triton-X and 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together