Dengue envelope-2 22 kDa
Fournisseur: Biorbyt
Certificats
À propos de cet article
Spécifications
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:Dengue Envelope-2 22kDa
- Purity:Protein is >95% pure as determined by 10% PAGE (coomassie staining).
- Sequence:ATLRKYCIEAKLTNTTTDSRCPTQGEPSLNEEQDKRFVCKHSMADRGWGNGCGLFGKGGIVTCAMFTCKKNMK GKVVQPENLEYTIVITPHSGEEHAVGNDTGKHGKEIKITPQSSITEAELTGYGAVTMECSPRTGLDFNEMVLL QMENKAWLVHRQWFLDLP.
- Formulation:50mM sodium chloride-phosphorous buffer, pH-7.5.
- Tested applications:SDS-PAGE , HPLC , FuncS
Spécifications
Frequently Bought Together