HIV-type O envelope
Catalog # BIRBORB85288-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Storage conditions:Store at−20 °C.
- Protein/peptide name:HIV-type O Envelope
- Purity:Greater than 95% by SDS-PAGE and RP-HPLC
- Formulation:Liquid. Supplied in 20 mM PBS pH 7.8, 0.5 M NaCl,1 mM DTT, 8 M urea and 0.4 M imidazole.
- Tested applications:WB , SDS-PAGE