HIV-1 envelope
Catalogus # BIRBORB85310-100
Leverancier: Biorbyt
CAS-nummer:
Specificaties
- Pk:100 µG
- Conjugation:Unconjugated
- Storage conditions:Store at−20 °C.
- Biological activity:Immunoreactive with all sera of HIV-1 and HIV-type Oinfected individuals and with 60-80% of HIV-2 infectedindividuals.This antigen was thoroughly tested in various tests as wellas ELISA using the standard BBI serum panels.All antibody detection tests on this antigen were assensitive or more than the standard Abott third generation tests.
- Protein/peptide name:HIV-1 Envelope
- Purity:Greater than 98% by HPLC
- Formulation:Liquid. Supplied in 0.5x PBS containing 6M urea.
- Tested applications:WB , SDS-PAGE