Human recombinant VHL (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Avoid multiple freeze-thaw cycles.
- Protein synonyms:Von Hippel-Lindau disease tumor suppressor
- Protein/peptide name:VHL
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Sequence:MGSSHHHHHH SSGLVPRGSH MPRRAENWDE AEVGAEEAGV EEYGPEEDGG EESGAEESGPEESGPEELGA EEEMEAGRPR PVLRSVNSRE PSQVIFCNRS PRVVLPVWLN FDGEPQPYPT LPPGTGRRIH SYRGHLWLFR DAGTHDGLLV NQTELFVPSL NVDGQPIFAN ITLP.
- Formulation:The Von Hippel-Lindau Protein contains 1x PBS pH-7.4, 2mM EDTA, and 1mM DTT.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together