Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant EBAG9 (from E. coli)
Human Recombinant EBAG9 (from E. coli)
Supplier:  Biorbyt
Certificates
Human Recombinant EBAG9 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    EBAG9 although stable 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
  • Protein synonyms:
    EB9
  • Protein/peptide name:
    EBAG9
  • Purity:
    Greater than 90% as determined by SDS-PAGE.
  • Sequence:
    GSSHHHHHH SSGLVPRGSH MRSGRGRKLS GDQITLPTTV DYSSVPKQTD VEEWTSWDED APTSVKIEGG NGNVATQQNS LEQLEPDYFK DMTPTIRKTQ KIVIKKREPL NFGIPDGSTG FSSRLAATQD LPFIHQSSEL GDLDTWQENT NAWEEEEDAA WQAEEVLRQQ KLADREKRAA EQQRKKMEKE AQRLMKKEQN KIGVKLS.
  • Formulation:
    The EBAG9 protein solution contains 20 mM Tris-HCl pH-8, 1mM DTT, 2mM EDTA and 10% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together