Order Entry
Belgium
ContactUsLinkComponent
Human recombinant PTPN1 (from E. coli)
Human recombinant PTPN1 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant PTPN1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Avoid multiple freeze-thaw cycles.
  • Biological activity:
    10,000 U/mg.
  • Protein synonyms:
    Tyrosine-protein phosphatase non-receptor type 1
  • Protein/peptide name:
    PTPN1
  • Purity:
    Greater than 95.0% as determined by(a)Analysis by RP-HPLC.(b)Analysis by SDS-PAGE.
  • Sequence:
    memekefeqi dksgswaaiy qdirheasdf pcrvaklpkn knrnryrdvs pfdhsriklhqedndyinas likmeeaqrs yiltqgplpn tcghfwemvw eqksrgvvml nrvmekgslkcaqywpqkee kemifedtnl kltlisedik syytvrqlel enlttqetre ilhfhyttwpdfgvpespas flnflfkvre sgslspehgp vvvhcsagig rsgtfcladt clllmdkrkdpssvdikkvl lemrkfrmgl iqtadqlrfs ylaviegakf imgdssvqdq wkelshedle pppehipppprppkrilephn.
  • Formulation:
    The protein contains 25mM Tris-HCl, pH 7.5, 2mM b-mercaptoethanol, 1mM EDTA, 1mMDTT and 20% Glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together