Human recombinant IL1RA (fromE. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze/thaw cycles.
- Protein synonyms:DIRA
- Protein/peptide name:IL1RA
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Formulation:Interleukin-1Ra His-tag protein is supplied in 20 mM Tris HCl (pH 8) and 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together