Rat S100 calcium binding G (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Rat
- Storage conditions:Lyophilized CABP9K although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution CABP9K should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:Protein S100-G
- Protein/peptide name:S100 calcium binding G
- Purity:Greater than 90% as determined by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized CABP9K in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:The protein was lyophilized from concentrated solution (1mg/ml) containing 100mM Phosphate buffer, pH 7.3.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together