Order Entry
Belgium
ContactUsLinkComponent
PKACa2 (from E. coli)
PKACa2 (from E. coli)
Supplier:  Biorbyt
Certificates
PKACa2 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Storage conditions:
    PKA should be stored at 4 °C if entire vial will be used within 2-4 weeks. For long term storage it is recommended to store at −20 °C. Avoid multiple freeze-thaw cycles.
  • Biological activity:
    Holoenzyme can be activated by adding the second messenger cAMP (Activation constant about 100nM) releasing two monomeric catalytic subunits.
  • Protein synonyms:
    Protein Kinase holoenzyme type II alpha
  • Protein/peptide name:
    PKACa2
  • Purity:
    Greater than 95% as determined by SDS-PAGE.
  • Formulation:
    PKA holoenzyme type-II alpha is supplied in 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together