Protein G (from E. coli)
Catalog # BIRBORB82629-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 g
- Source:E. coli
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:Less than 0.1 EU/ug of Protein G as determined by LAL method.
- Protein/peptide name:Protein G
- Purity:>95% pure by SDS-PAGE analyses.
- Molecular weight:Predicted at 21.6 kDa, but showed a band of 32 kDa in SDS-PAGE.
- Formulation:Lyophilized with no additive. Dissolve in distilled water or saline.
- Tested applications:SDS-PAGE , HPLC , FuncS