Human recombinant Collagen IX (from cells)
Catalog # BIRBORB138605-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Protein synonyms:Collagen alpha 1(IX) chain
- UniProtKB:P20849-3
- Protein/peptide name:Collagen IX
- Sequence:Met1-Pro328