Order Entry
Belgium
ContactUsLinkComponent
 
Human I-309 (from E. coli)
 :  Biorbyt
 
Human I-309 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein/peptide name:
    I-309
  • Purity:
    > 97% by SDS-PAGE and HPLC analyses
  • Molecular weight:
    8.5 kDa
  • Sequence:
    SKSMQVPFSR CCFSFAEQEI PLRAILCYRN TSSICSNEGL IFKLKRGKEACALDTVGWVQ RHRKMLRHCP SKRK
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in 20mM PB, pH 7.4, 100mM NaCl
  • Tested applications:
    SDS-PAGE , HPLC

 

 

Frequently Bought Together