Human TWEAK receptor (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:TNF-related weak inducer of apoptosis receptor
- Protein/peptide name:TWEAK Receptor
- Purity:> 95% by SDS-PAGE and HPLC analyses.
- Molecular weight:5.6 kDa
- Sequence:EQAPGTAPCS RGSSWSADLD KCMDCASCRA RPHSDFCLGC AAAPPAPFRL LWP
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together