Mouse SHH (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Mouse
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Murine Sonic Hedgehog
- Protein/peptide name:SHH
- Purity:> 95 % by SDS-PAGE and HPLC analyses
- Molecular weight:19.8 kDa
- Sequence:IVIGPGRGFG KRRHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKITRNSERFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVKLRVTEGWDED GHHSEESLHY EGRAVDITTS DRDRSKYGML ARLAVEAGFDWVYYESKAHI HCSVKAENSV AAKSGG
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together