Order Entry
Belgium
ContactUsLinkComponent
 
IL5 (from E. coli)
 :  Biorbyt
 
IL5 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein/peptide name:
    IL5
  • Tested applications:
    SDS-PAGE , HPLC

 

 

Frequently Bought Together