Order Entry
Belgium
ContactUsLinkComponent
Rat CINC-2 alpha (from E. coli)
Rat CINC-2 alpha (from E. coli)
Supplier:  Biorbyt
Certificates
Rat CINC-2 alpha (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    GRO-gamma
  • Protein/peptide name:
    CINC-2 alpha
  • Purity:
    > 95% by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    7.8 kDa
  • Sequence:
    RELRCQCLKT LPRVDFENIQ SLTVTPPGPH CTQTEVIATL KDGQEVCLNP QAPRLQKIIQKLLKSDKSS
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in 20mM PB, pH 7.4, 50mM NaCl.
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together