LcKGST SH3 domain
Catalog # BIRBORB98115-50
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:50 µG
- Storage conditions:Store at−80 °C.
- UniProtKB:1H92A
- Protein/peptide name:LcKGST SH3 domain
- Purity:90% pure by SDS-PAGE
- Formulation:Lyophilized