Order Entry
Belgium
ContactUsLinkComponent
HSV-II Ag (from E. coli)
HSV-II Ag (from E. coli)
Catalog # BIRBORB98962-1
Supplier:  Biorbyt
CAS Number:  
HSV-II Ag (from E. coli)
Catalog # BIRBORB98962-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
    E. coli
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/peptide name:
    HSV-II Ag
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulation:
    PBS, pH 7.4
  • Tested applications:
    ELISA , WB , IHC-P , RIA

Specifications

About this item