Recombinant CRP Ag
BIRBORB98923-1
: Biorbyt
:
- Pk:1 mg
- Protein/peptide type:Recombinant
- Storage conditions:Store at −20 °C. Avoid multiple freeze/thaw cycles.
- Protein/peptide name:CRP Ag
- Purity:Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
- Formulation:PBS, pH 7.4
- Tested applications:ELISA