Order Entry
Belgium
ContactUsLinkComponent
Recombinant cTnT Ag
Recombinant cTnT Ag
Catalog # BIRBORB98918-100
Supplier:  Biorbyt
CAS Number:  
Recombinant cTnT Ag
Catalog # BIRBORB98918-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/peptide name:
    cTnT Ag
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulation:
    PBS, pH 7.4
  • Tested applications:
    ELISA

Specifications

About this item