Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mouse Pre-S1 Ag

Mouse Pre-S1 Ag

1 mg
Catalog # BIRBORB98864-1
Marketsource
Supplier:   Biorbyt
Mouse Pre-S1 Ag
Catalog # BIRBORB98864-1
Supplier:  Biorbyt
Marketsource
CAS Number:  
Mouse Pre-S1 Ag

1 mg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 mg
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    BIRBORB98864-1
  • Protein/peptide name:
    Pre-S1 Ag
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Formulation:
    PBS, pH 7.4
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Tested applications:
    ELISA , WB , IHC-P , RIA
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.