Recombinant GPC3Ag
Catalog # BIRBORB98859-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Storage conditions:Store at −20 °C. Avoid multiple freeze/thaw cycles.
- Protein/peptide name:GPC3Ag
- Purity:Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
- Formulation:PBS, pH 7.4
- Tested applications:ELISA