Order Entry
Belgium
ContactUsLinkComponent
 
Cyfra211 Ag (from Cells)
  BIRBORB98830-100
 :  Biorbyt
 :  
Cyfra211 Ag (from Cells)
  BIRBORB98830-100
 :  Biorbyt
 :  

 

  • Pk:
    100 µG
  • Source:
    Cells
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/peptide name:
    Cyfra211 Ag
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulation:
    PBS, pH 7.4
  • Tested applications:
    ELISA