Order Entry
Belgium
ContactUsLinkComponent
beta-HCG Ag
beta-HCG Ag
Catalogus # BIRBORB98823-0.5
Leverancier:  Biorbyt
CAS-nummer:  
beta-HCG Ag
Catalogus # BIRBORB98823-0.5
Leverancier:  Biorbyt
CAS-nummer:  

Specificaties

  • Pk:
    0,5 mg
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/peptide name:
    beta-HCG Ag
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulation:
    PBS, pH 7.4
  • Tested applications:
    ELISA

Specificaties

Over dit artikel