Respiratory Syncytial Virus
Catalog # BIRBORB79786-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Storage conditions:Store at −20 °C.
- Protein/peptide name:Respiratory Syncytial Virus
- Purity:Greater than 95% by SDS-PAGE gel analysis.