Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Parainfluenza Virus Type 2
Parainfluenza Virus Type 2
Parainfluenza Virus Type 2
BIRBORB79784-1
:
Biorbyt
:
Parainfluenza Virus Type 2
BIRBORB79784-1
:
Biorbyt
:
Pk:
1 mg
Storage conditions:
Store at −20 °C.
Protein/peptide name:
Parainfluenza Virus Type 2
Purity:
Greater than 95% by SDS-PAGE gel analysis.