Order Entry
Belgium
ContactUsLinkComponent
H3N2 strain A/Shangdong/9/93
H3N2 strain A/Shangdong/9/93
Catalog # BIRBORB79773-1
Supplier:  Biorbyt
CAS Number:  
H3N2 strain A/Shangdong/9/93
Catalog # BIRBORB79773-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Storage conditions:
    Store at −20 °C.
  • Protein/peptide name:
    H3N2 strain A/Shangdong/9/93
  • Purity:
    Greater than 95% by SDS-PAGE gel analysis.

Specifications

About this item