Order Entry
Belgium
ContactUsLinkComponent
Rabbit Troponin I (from Skeletal muscle)
Rabbit Troponin I (from Skeletal muscle)
Catalog # BIRBORB79715-0.1
Supplier:  Biorbyt
CAS Number:  
Rabbit Troponin I (from Skeletal muscle)
Catalog # BIRBORB79715-0.1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    0,1 mg
  • Source:
    Skeletal muscle
  • Species:
    Rabbit
  • Storage conditions:
    Store at -2 to -8 °C.
  • Protein/peptide name:
    Troponin I
  • Purity:
    Greater than 95% pure SDS-PAGE. Protein G Chromatography

Specifications

About this item