Troponin troponin I (from skeletal muscle)
Catalog # BIRBORB79712-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Source:Skeletal muscle
- Species:Troponin
- Storage conditions:Store at -2 to -8 °C.
- Protein/peptide name:Troponin I
- Purity:Greater than 95% pure SDS-PAGE. Protein G Chromatography