Human recombinant recombinant glycosylated proBNP
Catalog # BIRBORB79667-0.1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:0,1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Storage conditions:Store at -2 to -8 °C.
- Protein/peptide name:Recombinant Glycosylated proBNP
- Purity:Greater than 95% pure SDS-PAGE. Protein G Chromatography