Human recombinant IGFBP6
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Storage conditions:The protein is stable at −20 °C. to −70 °C. in a manual defrost freezer.Avoid repeated freeze thaw cycles
- Protein/peptide name:IGFBP6
- Purity:Greater than 95% by SDS-PAGE gel analysis.
- Formulation:PBS, pH 7.2, sterile filtered
- Tested applications:ELISA , WB , DOT , FuncS
Specifications
Frequently Bought Together