Order Entry
Belgium
ContactUsLinkComponent
Human recombinant CDKN1A1 (from E. coli)
Human recombinant CDKN1A1 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant CDKN1A1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CyclinDependent Kinase Inhibitor 1
  • UniProtKB:
    P38936 (Human)
  • Protein/peptide name:
    CDKN1A1
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    SEPAGDVRQNPCGSKACRRLFGPVDSEQLRRDCDALMAGCIQEARERWNFDFVTETPLEGDFAWE RVRGLGLPKLYLPTGPRRGRDELGGGRRPGTSPALLQGTAEEDHVDLSLSCTLVPRSGEQAEGSP GGPGDSQGRKRRQTSMTDFYHSKRRLIFSKRKPLEHHHHHH
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together