Human recombinant urokinase (from HEK293 cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:UrokinaseType Plasminogen Activator
- UniProtKB:P00749 (Human)
- Protein/peptide name:Urokinase
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:SNELHQVPSNCDCLNGGTCVSNKYFSNIHWCNCPKKFGGQHCEIDKSKTCYEGNGHFYRGKASTD TMGRPCLPWNSATVLQQTYHAHRSDALQLGLGKHNYCRNPDNRRRPWCYVQVGLKPLVQECMVHD CADGKKPSSPPEELKFQCGQKTLRPRFKIIGGEFTTIENQPWFAAIYRRHRGGSVTYVCGGSLIS PCWVISATHCFIDYPKKEDYIVYLGRSRLNSNTQGEMKFEVENLILHKDYSADTLAHHNDIALLK IRSKEGRCAQPSRTIQTICLPSMYNDPQFGTSCEITGFGKENSTDYLYPEQLKMTVVKLISHREC QQPHYYGSEVTTKMLCAADPQWKTDSCQGDSGGPLVCSLQGRMTLTGIVSWGRGCALKDKPGVYT RVSHFLPWIRSHTKEENGLALVDHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM HEPES, 150mM NaCl, 2mM CaCl, 10% Glycerol, pH 7.5.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together