Human recombinant OTUB2 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Ubiquitin Thioesterase OTUB2
- UniProtKB:Q96DC9 (Human)
- Protein/peptide name:OTUB2
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MSPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKL TQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEML KMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKS SKYIAWPLQGWQATFGGGDHPPKSDLVPRGSHMSETSFNLISEKCDILSILRDHPENRIYRRKIE ELSKRFTAIRKTKGDGNCFYRALGYSYLESLLGKSREIFKFKERVLQTPNDLLAAGFEEHKFRNF FNAFYSVVELVEKDGSVSSLLKVFNDQSASDHIVQFLRLLTSAFIRNRADFFRHFIDEEMDIKDF CTHEVEPMATECDHIQITALSQALSIALQVEYVDEMDTALNHHVFPEAATPSVYLLYKTSHYNIL YAADKH
- Formulation:Supplied as a 0.2 um filtered solution of 50mM HEPES, 150mM NaCl, 2mM DTT, pH 7.5 .
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together