Human recombinant TXNDC12 (from HEK293 cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Thioredoxin DomainContaining 12
- UniProtKB:O95881 (Human)
- Protein/peptide name:TXNDC12
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:HNGLGKGFGDHIHWRTLEDGKKEAAASGLPLMVIIHKSWCGACKALKPKFAESTEISELSHNFVM VNLEDEEEPKDEDFSPDGGYIPRILFLDPSGKVHPEIINENGNPSYKYFYVSAEQVVQGMKEAQE RLTGDAFRKKHLVDHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 10% Glycerol, pH 7.4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together