Order Entry
Belgium
ContactUsLinkComponent
Human recombinant BMP2 (from E. coli)
Human recombinant BMP2 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant BMP2 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Biological activity:
    Recombinant BMP2 is fully biologically active when compared to standards.
    ED50 is less than 50 ng/ml as determined by the cytolysis of MC3T3E1 cells.
    Specific Activity of 2.0 x 104 IU/mg
  • Protein synonyms:
    Bone Morphogenetic 2
  • UniProtKB:
    P12643 (Human)
  • Protein/peptide name:
    BMP2
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MQAKHKQRKRLKSSCKRHPLYVDFSDVGWNDWIVAPPGYHAFYCHGECPFPLADHLNSTNHAIVQ TLVNSVNSKIPKACCVPTELSAISMLYLDENEKVVLKNYQDMVVEGCGCR
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 50mM HAc .
  • Tested applications:
    SDS-PAGE , FuncS

Specifications

Related Information

Frequently Bought Together