Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant BCL2L1 (from E. coli)
 :  Biorbyt
 
Human recombinant BCL2L1 (from E. coli)
 :  Biorbyt

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Apoptosis regulator Bcl X
  • UniProtKB:
    Q07817 (Human)
  • Protein/peptide name:
    BCL2L1
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MSQSNRELVVDFLSYKLSQKGYSWSQFSDVEENRTEAPEGTESEMETPSAINGNPSWHLADSPAV NGATGHSSSLDAREVIPMAAVKQALREAGDEFELRYRRAFSDLTSQLHITPGTAYQSFEQVVNEL FRDGVNWGRIVAFFSFGGALCVESVDKEMQVLVSRIAAWMATYLNDHLEPWIQENGGWDTFVELY GNNAAAESRKGQERFNRLEHHHHHH
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM HEPES, 50mM KCl, 20% Glycerol, pH 7.5.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together