Human recombinant Annexin A13 (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Annexin A13|Annexin-13|ANXA13|Intestine-Specific Annexin|ISA|ANX13|Annexin XIII
- UniProtKB:P27216 (Human)
- Protein/peptide name:Annexin A13
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:GNRHAKASSPQGFDVDRDAKKLNKACKGMGTNEAAIIEILSGRTSDERQQIKQKYKATYGKELEE VLKSELSGNFEKTALALLDHPSEYAARQLQKAMKGLGTDESVLIEVLCTRTNKEIIAIKEAYQRL FDRSLESDVKGDTSGNLKKILVSLLQANRNEGDDVDKDLAGQDAKDLYDAGEGRWGTDELAFNEV LAKRSYKQLRATFQAYQILIGKDIEEAIEEETSGDLQKAYLTLVRCAQDCEDYFAERLYKSMKGA GTDEETLIRIIVTRAEVDLQGIKAKFQEKYQKSLSDMVRSDTSGDFRKLLVALLH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 10% Glycerol, pH 8.0.
- Tested applications:SDS-PAGE
Frequently Bought Together