Order Entry
Belgium
ContactUsLinkComponent
Recombinant T.pallidum p15 (from bacteria)
Recombinant T.pallidum p15 (from bacteria)
Catalog # BIRBORB48260-1
Supplier:  Biorbyt
CAS Number:  
Recombinant T.pallidum p15 (from bacteria)
Catalog # BIRBORB48260-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    Bacteria
  • Storage conditions:
    Upon receipt, store at to−20 °C.Avoid multiple/freeze thaw cycles.
  • Protein/peptide name:
    T.pallidum p15
  • Purity:
    Nickel-NTA affinity chromatography. Major band at 19 kDa, minor band at 15 kDa on 12% PAGECoomassie Staining.
  • Formulation:
    150 mM Imidazole, 25 mM Sodium Phosphate, pH 8.0 containing 150 mM Sodium Chloride, 50% Glycerol
  • Tested applications:
    ELISA

Specifications

About this item