Order Entry
Belgium
ContactUsLinkComponent
Recombinant IFN beta 1b
Recombinant IFN beta 1b
Catalog # BIRBORB97782-10
Supplier:  Biorbyt
CAS Number:  
Recombinant IFN beta 1b
Catalog # BIRBORB97782-10
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at−20 °C.
  • Biological activity:
    EC50: 3.2x107 IU/mg, determined in a viral resistanceassay human "Wish" cell line and VSV virus or themonkey VERO cell line with EMCV virus.
  • Protein synonyms:
    beta-interferon
  • UniProtKB:
    P01574
  • Protein/peptide name:
    IFN beta 1b
  • Purity:
    95% pure by SDS-PAGE and RP-HPLC
  • Solubility:
    It is recommended to reconstitute the lyophilized IFN- insterile bidest water not less than 100 g/ml, which canthen be further diluted to other aqueous solutions
  • Molecular weight:
    19 kDa
  • Formulation:
    Lyophilized. Lyophilized from a 1mg/ml solution containing 50mg human albumine and 50mg dextrose

Specifications

About this item