Order Entry
Belgium
Orders LinkContactUsLinkComponent
GH1

GH1

500 µG
Catalog # BIRBORB97768-500
Marketsource
Supplier:   Biorbyt
GH1
Catalog # BIRBORB97768-500
Supplier:  Biorbyt
Marketsource
CAS Number:  
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 µG
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    500 µG
  • Cat. No.:
    BIRBORB97768-500
  • Protein/peptide name:
    GH1
  • Storage conditions:
    Store at−20 °C.
  • Formulation:
    Lyophilized. Pituitary Growth Hormone protein waslyophilized from a 1 mg/ml solution with 0.0045 mMNaHCO3 and pH 11.0.
  • Solubility:
    It is recommended to reconstitute and dilute the lyophilizedGH in 0.4% NaHCO3 adjusted to pH 11.0, which canthen be further diluted to other aqueous solutions. It isrecommended when diluting to below 10 ug/ml that thedilution solution will contain carrier
  • UniProtKB:
    P01241
  • Purity:
    95% pure by SDS-PAGE and RP-HPLC
  • Biological activity:
    3 U/mg
  • Molecular weight:
    22 kDa

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.